missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gallery Reagents for Food and Beverage Analysis

Product Code. 17914041 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Alpha Amylase System Reagent
Alpha to Amino Nitrogen
Sucrose System Reagent
Quantity:
2 Reagents in 20 mL Vials
20 mL Vials
Reagent A: 6 x 20 mL Vials
Reagent B: 2 x 5 mL Vials
Unit Size:
200 tests
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity Unit Size
17914041 Alpha Amylase System Reagent Reagent A: 6 x 20 mL Vials
Reagent B: 2 x 5 mL Vials
200 tests
17861911 Sucrose System Reagent 20 mL Vials Each
17717697 Alpha to Amino Nitrogen 2 Reagents in 20 mL Vials Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17914041 Supplier Thermo Scientific™ Supplier No. 984373
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Choose from a continuously expanding selection of tests for automated food and beverage analysis.

Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for food and beverage analysis. Over 50 ready-to-use gallery system reagents are offered for food and beverage analysis, with the kits including two to four reagents in 20 mL barcoded vials. This enables the necessary identification data, such as lot numbers and expiration dates, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations. Save time and reduce errors by simplifying your analysis with the Gallery system reagents.

  • Kit structure includes 2 to 4 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, is automatically read
  • Photometric measurement is mainly based on the formation or consumption of NAD or NADH
  • Measurements are done at 340nm in 37°C
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
TRUSTED_SUSTAINABILITY

Specifications

Type Alpha Amylase System Reagent
Sample Type Food and Beverage Sample
Description Alpha-Amylase system reagents (200 tests)
Detects Alpha-Amylase
No. of Tests 200
Test Range 10 to 7500 U/I
Analytical Method Enzymatic test with ethylidene-pNP-G7 (E-pNP-G7) as the substrate
Quantity Reagent A: 6 x 20 mL Vials
Reagent B: 2 x 5 mL Vials
Interference Filter 405
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.