Learn More
Thermo Scientific™ Gallery™ System Reagents for Environmental and Industrial Analysis
Shop All Thermo Scientific ProductsDescription
Select from these ready-to-use reagents and enzymatic kits specifically designed for environmental and industrial water analysis using a Thermo Scientific™ Gallery™ Discrete Analyzer.
Save time and reduce errors by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use Gallery system reagents, these kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the Gallery Discrete Analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries.
- All test applications are based on recommended standard methods in water and wastewater analysis of different countries
- Kit structure includes 1 to 3 reagents in 20mL vials
- Barcoded vials are simple and fast to use
- All necessary data, such as lot and expiration date, are automatically read
- Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
- Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
Specifications
Specifications
| Type | Phosphate System Reagent |
| Sample Type | Water Sample, Soil Sample |
| Description | Phosphate system reagents - R1 (2300 tests) |
| Detects | Phosphate R1 |
| No. of Tests | 2300 |
| Test Range | 1.0 to 10.0 |
| Quantity | 4 x 20 mL |
| Interference Filter | 880 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.