missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gallery™ System Reagents for Environmental and Industrial Analysis

Product Code. 17828533 Shop All Thermo Scientific Products
Click to view available options
Type:
Fluoride System Reagent
Fluoride System Reagent R1
Fluoride System Reagent R3
Phosphate System Reagent
No. of Tests:
2200
2300
3000
4800
Unit Size:
1 set
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 17828533

Brand: Thermo Scientific™ 984630

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Select from these ready-to-use reagents and enzymatic kits specifically designed for environmental and industrial water analysis using a Thermo Scientific™ Gallery™ Discrete Analyzer.

Save time and reduce errors by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use Gallery system reagents, these kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the Gallery Discrete Analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries.

Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water

Specifications

Type Fluoride System Reagent
Sample Type Water Sample, Soil Sample
Description Fluoride system reagents - R2 (4800 tests)
Detects Fluoride R2
No. of Tests 4800
Test Range 0 to 5.0 (for Gallery), 0 to 2.0 (for Aquakem)
Quantity 4 x 20 mL
Interference Filter 600/ 880
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.