Learn More
Thermo Scientific™ Gallery™ System Reagents for Environmental and Industrial Analysis
Product Code. 17818533
Change view
Click to view available options
Type:
Fluoride System Reagent
Fluoride System Reagent R1
Fluoride System Reagent R3
Phosphate System Reagent
No. of Tests:
2200
2300
3000
4800
Unit Size:
1 set
Each
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type No. of Tests unitSize
17818533 Fluoride System Reagent R1 2200 1 set
17453333 Phosphate System Reagent 2300 Pack of 4
17463333 Phosphate System Reagent 3000 Each
17828533 Fluoride System Reagent 4800 1 set
17838533 Fluoride System Reagent R3 2200 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17818533 Supplier Thermo Scientific™ Supplier No. 984629
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Select from these ready-to-use reagents and enzymatic kits specifically designed for environmental and industrial water analysis using a Thermo Scientific™ Gallery™ Discrete Analyzer.
Save time and reduce errors by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use Gallery system reagents, these kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the Gallery Discrete Analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries.
Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
Specifications
Type Fluoride System Reagent R1
Sample Type Water Sample, Soil Sample
Description Fluoride system reagents - R1 (2200 tests)
Detects Fluoride R1
No. of Tests 2200
Test Range 0 to 5.0 (for Gallery), 0 to 2.0 (for Aquakem)
Quantity 4 x 20 mL
Interference Filter 600/ 880