missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gallery™ System Reagents for Environmental and Industrial Analysis

Product Code. 17453333 Shop All Thermo Scientific Products
Click to view available options
Type:
Fluoride System Reagent
Fluoride System Reagent R1
Fluoride System Reagent R3
Phosphate System Reagent
No. of Tests:
2200
2300
3000
4800
Unit Size:
1 set
Each
Pack of 4
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 17453333

Brand: Thermo Scientific™ 984366

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Select from these ready-to-use reagents and enzymatic kits specifically designed for environmental and industrial water analysis using a Thermo Scientific™ Gallery™ Discrete Analyzer.

Save time and reduce errors by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use Gallery system reagents, these kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the Gallery Discrete Analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries.

Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
TRUSTED_SUSTAINABILITY

Specifications

Type Phosphate System Reagent
Sample Type Water Sample, Soil Sample
Description Phosphate system reagents - R1 (2300 tests)
Detects Phosphate R1
No. of Tests 2300
Test Range 1.0 to 10.0
Quantity 4 x 20 mL
Interference Filter 880
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.