missing translation for 'onlineSavingsMsg'
Learn More

Gallamine Triethiodide, MedChemExpress

Product Code. 16452126
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452126 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452126 Supplier Medchem Express Supplier No. HYB0416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gallamine Triethiodide is a synthetic nondepolarizing blocking drug.

Gallamine Triethiodide is a synthetic nondepolarizing blocking drug.

Specifications

Chemical Name or Material Gallamine Triethiodide
CAS 65-29-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H60I3N3O3
Quantity 500 mg
Solubility Information H2O : ≥ 30 mg/mL (33.65 mM)
SMILES CC[N+](CC)(CC)CCOC(C(OCC[N+](CC)(CC)CC)=CC=C1)=C1OCC[N+](CC)(CC)CC.[I-].[I-].[I-]
Molecular Weight (g/mol) 891.53
Formula Weight 891.53
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.