Learn More
Invitrogen™ GALK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534699
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (89%), Rat (89%), Chicken (81%), Bovine (93%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a highly efficient N-acetylgalactosamine kinase, which has galactokinase activity when galactose is present at high concentrations. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| GALK2 | |
| Polyclonal | |
| Unconjugated | |
| GALK2 | |
| 2810017M24Rik; AI327030; Galactokinase 2; GALK2; GalNAc kinase; Gk2; N-acetylgalactosamine kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2585 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q01415 | |
| GALK2 | |
| Recombinant protein encompassing a sequence within the center region of human GALK2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.