missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-9 Mouse anti-Human, PE, Clone: 9M1-3, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565892
This item is not returnable.
View return policy
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
| Galectin-9 | |
| Monoclonal | |
| PE | |
| galectin 9; Gal-9; Ecalectin; HUAT; LGALS9; LEG9; Tumor antigen HOM-HD-21 | |
| Human Galectin-9 (AA 208-215) Peptide | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
| IgG1 κ |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| 9M1-3 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction