missing translation for 'onlineSavingsMsg'
Learn More

Galectin-9 Antibody (B-T33) - Azide and BSA Free, Novus Biologicals™

Product Code. 18371335 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371335 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371335 Supplier Novus Biologicals Supplier No. NBP318123

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for ELISA (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-9
Applications ELISA
Classification Monoclonal
Clone B-T33
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 1-10ug/ml
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Host Species Mouse
Immunogen Recombinant human Galectin-9. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3965
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.