missing translation for 'onlineSavingsMsg'
Learn More

Galectin-9 Antibody (9M1-3), DyLight 405, Novus Biologicals™

Product Code. 18387915 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387915 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387915 Supplier Novus Biologicals Supplier No. NBP318530V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-9
Applications Flow Cytometry, Immunohistochemistry, Flow Cytometry
Classification Monoclonal
Clone 9M1-3
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Host Species Mouse
Immunogen Recombinant M-type splicing variant of human Galectin-9
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3965
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.