missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galectin 8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA528506

Product Code. 13492617

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T transfected recombinant LGALS8 protein. Predicted reactivity: Rat (80%), Dog (86%), Pig (83%), Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the galectin family. Galectins are beta-galactoside-binding animal lectins with conserved carbohydrate recognition domains. The galectins have been implicated in many essential functions including development, differentiation, cell-cell adhesion, cell-matrix interaction, growth regulation, apoptosis, and RNA splicing. This gene is widely expressed in tumoral tissues and seems to be involved in integrin-like cell interactions. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Galectin 8
Polyclonal
Unconjugated
Lgals8
1200015E08Rik; 30 kDa S-type lectin; AI326142; D13Ertd524e; Gal-8; galectin 8; galectin-8; galectin-8g; galectin-8L; lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 8; lectin, galactoside binding soluble 8; lectin, galactoside-binding, soluble, 8; LGALS8; Lgals-8; PCTA1; PCTA-1; Po66 carbohydrate binding protein; po66 carbohydrate-binding protein; Po66-CBP; prostate carcinoma tumor antigen 1; RL-30; RP11-385F5.1
Rabbit
Antigen affinity chromatography
RUO
3964
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O00214
Lgals8
Recombinant protein encompassing a sequence within the center region of human Galectin 8. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.