missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549931
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total Galectin 7 protein.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19.Specifications
Galectin 7 | |
Polyclonal | |
Unconjugated | |
LGALS7 | |
GAL7; Gal-7; galectin 7; galectin 7B; galectin-7; galectin-7B; HKL-14; keratinocyte lectin 14; lectin galactoside-binding soluble 7; lectin, galactose binding, soluble 7; lectin, galactoside binding soluble 7; lectin, galactoside binding soluble 7B; lectin, galactoside-binding, soluble, 7; lectin, galactoside-binding, soluble, 7B; LGALS7; LGALS7A; LGALS7B; p53-induced gene 1 protein; p53-induced protein 1; PI7; Pig1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16858, 29518, 3963, 653499 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O54974, P47929, P97590 | |
LGALS7, LGALS7B | |
Synthesized peptide derived from internal of human LEG7. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |