missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145325
This item is not returnable.
View return policy
Description
Galectin 5 is a member of the galectin family of proteins, which are characterized by their ability to bind to specific carbohydrate structures on cell surfaces. Galectin 5 is primarily expressed in the cytoplasm of cells and has been shown to play a role in regulating cell growth, differentiation, and apoptosis. It has also been implicated in various physiological processes, including inflammation, immune response, and cancer progression. Galectin 5 has been found to interact with a variety of proteins and signaling pathways, suggesting that it may have multiple functions within cells.
Specifications
| Galectin 5 | |
| Polyclonal | |
| Unconjugated | |
| Lgals5 | |
| Lgals5 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 25475 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P47967 | |
| Lgals5 | |
| Recombinant Rat GAL5 protein, Met1-Thr145 (Accession #P47967). | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction