missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534836
This item is not returnable.
View return policy
Description
Recommended positive controls: LGALS4 transfected 293T lysate. Predicted reactivity: Rabbit (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. Galectin 4 binds lactose and a related range of sugars. May be involved in the assembly of adherens junctions.
Specifications
| Galectin 4 | |
| Polyclonal | |
| Unconjugated | |
| Lgals4 | |
| Antigen NY-CO-27; GAL4; gal-4; galectin 4; galectin-4; L-36; L-36 lactose-binding protein; L36LBI; L36LBP; lactose-binding lectin 4; Lectin galactose binding soluble 4 (Galectin-4); lectin, galactose binding, soluble 4; Lectin, galactose binding, soluble 4 (Galectin-4); lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; lectin, galactoside-binding, soluble, 4 (galectin 4); Lgals4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1.02 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P56470 | |
| Lgals4 | |
| Recombinant protein encompassing a sequence within the center region of human Galectin 4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction