missing translation for 'onlineSavingsMsg'
Learn More

Galectin-4 Antibody (MM0300-1D12), DyLight 594, Novus Biologicals™

Product Code. 18419695 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419695 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419695 Supplier Novus Biologicals Supplier No. NBP212323DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-4 Monoclonal antibody specifically detects Galectin-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0300-1D12
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4
Host Species Mouse
Immunogen Human recombinant Galectin-4
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3960
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.