missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Galectin-3BP/MAC-2BP/LGALS3BP Polyclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Galectin-3BP/MAC-2BP/LGALS3BP |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 306-585 of human Galectin-3BP/MAC-2BP/LGALS3BP (NP_005558.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?