missing translation for 'onlineSavingsMsg'
Learn More

Galectin-3BP/MAC-2BP/LGALS3BP Rabbit anti-Human, Mouse, Rat, Clone: S03-5B8, Novus Biologicals™

Product Code. 18356285 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356285 100 μg 100µL
18354204 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18356285 Supplier Novus Biologicals Supplier No. NBP315031100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Galectin-3BP/MAC-2BP/LGALS3BP Monoclonal antibody specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-3BP/MAC-2BP/LGALS3BP
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone S03-5B8
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunofluorescence 20 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Host Species Rabbit
Immunogen Recombinant protein of human Galectin-3BP/MAC-2BP/LGALS3BP (Uniprot#: Q08380)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3959
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.