missing translation for 'onlineSavingsMsg'
Learn More

Galectin-3BP/MAC-2BP/LGALS3BP Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£153.00 - £366.00

Specifications

Antigen Galectin-3BP/MAC-2BP/LGALS3BP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18042076
View Documents
Novus Biologicals
NBP3-03777-20UL
20 μL
£153.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18362146
View Documents
Novus Biologicals
NBP3-03777-100UL
100 μL
£366.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Galectin-3BP/MAC-2BP/LGALS3BP Polyclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Galectin-3BP/MAC-2BP/LGALS3BP
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
3959
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Human, Mouse
90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Recombinant fusion protein containing a sequence corresponding to amino acids 6-221 of human Galectin-3BP/MAC-2BP/LGALS3BP (NP_005558.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.