missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Antibody (OTI6B7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01160
This item is not returnable.
View return policy
Description
Galectin-3BP/MAC-2BP/LGALS3BP Monoclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Galectin-3BP/MAC-2BP/LGALS3BP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:200-500, Flow Cytometry 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin 1:150 | |
| LGALS3BP | |
| Human recombinant protein fragment corresponding to amino acids 19-300 of human LGALS3BP(NP_005558) produced in E.coil. | |
| Affinity Purified | |
| RUO | |
| 3959 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI6B7 | |
| Unconjugated | |
| 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K | |
| Mouse | |
| 65.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction