missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97200V
This item is not returnable.
View return policy
Description
Galectin-3BP/MAC-2BP/LGALS3BP Polyclonal antibody specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Galectin-3BP/MAC-2BP/LGALS3BP | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K | |
| Produced in rabbits immunized with E. coli-derived Human Galectin-3BP/MAC-2BP/LGALS3BP fragment. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 3959 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction