Learn More
Galectin 3 Monoclonal Antibody (eBioM3/38 (M3/38)), PE, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£97.25 - £263.00
Specifications
| Antigen | Galectin 3 |
|---|---|
| Clone | eBioM3/38 (M3/38) |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15238799
|
Invitrogen
12-5301-81 |
50 μg | |||||||
15248799
![]() |
Invitrogen™
12-5301-82 |
100 μg |
£185.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15258799
|
Invitrogen™
12-5301-83 |
200 μg |
£263.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The eBioM3/38 antibody reacts with mouse Galectin-3 (Mac-2). Galectins are a family of animal lectins which appear to exhibit a variety of biological functions. Galectin 3 (aka LGALS3, galactose-specific soluble lectin 3, Mac-2, L-29) is one of the more extensively studied members of this family and is a 30 kDa beta-galactoside-binding protein. Due to a C-terminal carbohydrate binding site, Galectin 3 is capable of binding IgE and mammalian cell surfaces only when homodimerized or homo-oligomerized. Galectin 3 is normally distributed in epithelia of many organs and various inflammatory cells, including macrophages, as well as dendritic cells and Kupffer cells. The expression of this lectin is up-regulated during inflammation, cell proliferation, cell differentiation and through trans-activation by viral proteins. This monoclonal antibody eBioM3/38 crossreacts to human Galectin-3 (LGALS3). Applications Reported: This eBioM3/38 (M3/38) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioM3/38 (M3/38) antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal macrophages. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Galactose-specific lectin which binds IgE. May mediate with the alpha-3, beta-1 integrin the stimulation by CSPG4 of endothelial cells migration. Together with DMBT1, required for terminal differentiation of columnar epithelial cells during early embryogenesis. In the nucleus: acts as a pre-mRNA splicing factor. Involved in acute inflammatory responses including neutrophil activation and adhesion, chemoattraction of monocytes macrophages, opsonization of apoptotic neutrophils, and activation of mast cells.Specifications
| Galectin 3 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 16854, 3958 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| eBioM3/38 (M3/38) | |
| Flow Cytometry | |
| PE | |
| Rat | |
| Human, Mouse | |
| LGALS3 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| LGALS3 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.