missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3 Antibody (A3A12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£169.00
Specifications
| Antigen | Galectin-3 |
|---|---|
| Clone | A3A12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 6 μg/mL, Immunohistochemistry-Frozen 6 μg/mL |
| Applications | Western Blot |
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Galectin-3 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Markers, Tumor Suppressors | |
| P17931 | |
| 3958 | |
| Full length recombinant human galectin-3. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 26 kDa |
| A3A12 | |
| Western Blot 1 μg/mL, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 6 μg/mL, Immunohistochemistry-Frozen 6 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 | |
| LGALS3 | |
| IgG1 | |
| Immunogen affinity purified | |
| Galectin 3 (A3A12) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title