missing translation for 'onlineSavingsMsg'
Learn More

Galectin-3 Antibody (024), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18339800 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339800 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339800 Supplier Novus Biologicals Supplier No. NBP289383JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-3
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 24
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM Sodium Borate
Gene Alias Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Galectin-3 (P17931; Met1-Ile250).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3958
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.