missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 2 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149235
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction.
Specifications
| Galectin 2 | |
| Polyclonal | |
| Unconjugated | |
| LGALS2 | |
| 2200008F12Rik; AI324147; Beta-galactoside-binding lectin L-14-II; gal-2; galectin 2; galectin-2; HL14; lactose-binding lectin 2; lectin galactoside-binding soluble 2 (galectin 2); lectin, galactose-binding, soluble 2; lectin, galactoside binding soluble 2; lectin, galactoside-binding, soluble, 2; lectin, galactoside-binding, soluble, 2 (galectin 2); LEG2; Lgals2; S-Lac lectin 2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 107753 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9CQW5 | |
| LGALS2 | |
| E. coli-derived recombinant mouse Galectin-2, Ser2-Glu130 (Gly36Val). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction