missing translation for 'onlineSavingsMsg'
Learn More

Galectin-2 Antibody (1H6), Novus Biologicals™

Product Code. 18343733 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343733 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343733 Supplier Novus Biologicals Supplier No. NBP311293

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-2 Monoclonal specifically detects Galectin-2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-2
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
Clone 1H6
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Beta-galactoside-binding lectin L-14-II, Gal-2, galectin 2, galectin-2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, MGC75071, S-Lac lectin 2
Host Species Mouse
Immunogen Recombinant human galectin-2
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3957
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.