missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-13 Mouse, Clone: PP13/1165, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Galectin-13 |
---|---|
Clone | PP13/1165 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950138
|
EnquireBio
29124-MSM5-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940138
|
EnquireBio
29124-MSM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Galectin-13 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
29124 | |
IgG2b | |
Protein A/G Purified | |
Cytoplasmic and Cell Surface | |
K-562 cells. Placenta or Spleen. |
PP13/1165 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_016882204 | |
Recombinant human Galectin-13 protein fragment (aa23-134) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
16kDa (monomer) ;32kDa (homodimer) |