missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 13 Antibody (PP13/1165), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53168-100UG
This item is not returnable.
View return policy
Description
Galectin 13 Monoclonal specifically detects Galectin 13 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Galectin 13 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page | |
| beta-galactoside-binding lectin, EC 3.1.1.5, gal-13, GAL13, galectin-13, lectin, galactoside-binding, soluble, 13, PLAC8galectin 13, Placental protein 13, Placental tissue protein 13, PP13galactoside-binding soluble lectin 13 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 29124 | |
| Store at 4C. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| PP13/1165 | |
| Unconjugated | |
| 1mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| LGALS13 | |
| Recombinant human Galectin-13 protein fragment (aa23-134) (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction