missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 13 Antibody (PP13/1165) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£489.00
Specifications
| Antigen | Galectin 13 |
|---|---|
| Clone | PP13/1165 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1 ug/ml, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Description
Galectin 13 Monoclonal specifically detects Galectin 13 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Galectin 13 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| beta-galactoside-binding lectin, EC 3.1.1.5, gal-13, GAL13, galectin-13, lectin, galactoside-binding, soluble, 13, PLAC8galectin 13, Placental protein 13, Placental tissue protein 13, PP13galactoside-binding soluble lectin 13 | |
| LGALS13 | |
| IgG2b | |
| Protein A or G purified |
| PP13/1165 | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with No Preservative | |
| 29124 | |
| Recombinant human Galectin-13 protein fragment (aa23-134) (exact sequence is proprietary) | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title