missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 13 Antibody (PP13/1164), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54397V
This item is not returnable.
View return policy
Description
Galectin 13 Monoclonal antibody specifically detects Galectin 13 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Galectin 13 | |
| Monoclonal | |
| DyLight 405 | |
| beta-galactoside-binding lectin, EC 3.1.1.5, gal-13, GAL13, galectin-13, lectin, galactoside-binding, soluble, 13, PLAC8galectin 13, Placental protein 13, Placental tissue protein 13, PP13galactoside-binding soluble lectin 13 | |
| Recombinant human Galectin 13 protein fragment (aa23-134) (Uniprot: Q9UHV8) | |
| 100 μg | |
| metabolism | |
| 29124 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PP13/1164 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction