missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 13 Antibody (PP13/1164), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54397CL1
This item is not returnable.
View return policy
Description
Galectin 13 Monoclonal antibody specifically detects Galectin 13 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Galectin 13 | |
| Monoclonal | |
| CoraFluor 1 | |
| beta-galactoside-binding lectin, EC 3.1.1.5, gal-13, GAL13, galectin-13, lectin, galactoside-binding, soluble, 13, PLAC8galectin 13, Placental protein 13, Placental tissue protein 13, PP13galactoside-binding soluble lectin 13 | |
| Recombinant human Galectin 13 protein fragment (aa23-134) (Uniprot: Q9UHV8) | |
| 0.1 mL | |
| metabolism | |
| 29124 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| PP13/1164 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction