missing translation for 'onlineSavingsMsg'
Learn More

Galectin 13 Antibody (PP13/1162), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18427872 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427872 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427872 Supplier Novus Biologicals Supplier No. NBP254389AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin 13 Monoclonal antibody specifically detects Galectin 13 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 13
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone PP13/1162
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias beta-galactoside-binding lectin, EC 3.1.1.5, gal-13, GAL13, galectin-13, lectin, galactoside-binding, soluble, 13, PLAC8galectin 13, Placental protein 13, Placental tissue protein 13, PP13galactoside-binding soluble lectin 13
Host Species Mouse
Immunogen Recombinant human Galectin 13 protein fragment (aa23-134) (Uniprot: Q9UHV8)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 29124
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.