missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25225-1-AP-150UL
This item is not returnable.
View return policy
Description
Regulates immune responses through the recognition of cell-surface glycans. Essential for the anergy and suppressive function of CD25-positive regulatory T-cells (Treg).Specifications
| Galectin 10 | |
| Polyclonal | |
| Unconjugated | |
| CLC | |
| Charcot Leyden crystal protein, CLC, Eosinophil lysophospholipase, Gal 10, Galectin 10, LGALS10, LPPL_HUMAN, Lysolecithin acylhydrolase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1178 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q05315 | |
| CLC | |
| Galectin 10 Fusion Protein Ag18569 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction