missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 1 Recombinant Rabbit Monoclonal Antibody (SAIC-25B-112)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547904
This item is not returnable.
View return policy
Description
Specificity: Recognises human LGALS1. Preparation: The KLH-conjugated peptide 'SFVLNLGK' derived from Galectin 1. Galectin 1 binds beta-galactoside and other complex carbohydrates and may regulate apoptosis, cell proliferation and cell differentiation.
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.
Specifications
| Galectin 1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P09382 | |
| LGALS1 | |
| The KLH-conjugated peptide 'SFVLNLGK' derived from Galectin 1. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Western Blot | |
| SAIC-25B-112 | |
| Unconjugated | |
| LGALS1 | |
| 14 kDa laminin-binding protein; 14 kDa lectin; AA410090; beta-galactoside binding protein; beta-galactoside-binding lectin; beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14kDa; DKFZp686E23103; GAL1; GAL-1; Galaptin; Galbp; galectin 1; galectin 1; LGALS1; Galectin1; galectin-1; galectin-1-like protein; GBP; HBL; H-Galectin-1; HLBP14; HPL; L14; L-14.5; Lactose-binding lectin 1; Lect14; Lectin galactoside-binding soluble 1; lectin, galactose binding, soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; lectin, galactoside-binding, soluble, 1 (galectin 1); Lgals1; LGAS1; OPG-1; pm12; Putative MAPK-activating protein PM12; RL 14.5; similar to galactose-binding lectin; S-Lac lectin 1; unnamed protein product | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3956 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction