Learn More
Invitrogen™ Galectin 1 Recombinant Rabbit Monoclonal Antibody (23GB6025), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553938
 View more versions of this product
Product Code. 30088078
  • £485.00 / 100µL
Description
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galectin 1
Recombinant Monoclonal
Unconjugated
LGALS1
14 kDa laminin-binding protein; 14 kDa lectin; AA410090; beta-galactoside binding protein; beta-galactoside-binding lectin; beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14kDa; DKFZp686E23103; GAL1; GAL-1; Galaptin; Galbp; galectin 1; galectin 1; LGALS1; Galectin1; galectin-1; galectin-1-like protein; GBP; HBL; H-Galectin-1; HLBP14; HPL; L14; L-14.5; Lactose-binding lectin 1; Lect14; Lectin galactoside-binding soluble 1; lectin, galactose binding, soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; lectin, galactoside-binding, soluble, 1 (galectin 1); Lgals1; LGAS1; OPG-1; pm12; Putative MAPK-activating protein PM12; RL 14.5; similar to galactose-binding lectin; S-Lac lectin 1; unnamed protein product
Rabbit
Affinity chromatography
RUO
16852, 3956, 56646
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB6025
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P09382, P11762, P16045
LGALS1
A synthesized peptide derived from human Galectin 1 (80-120AA).
100 μL
Primary
Human, Mouse, Rat