missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11858-1-AP-20UL
This item is not returnable.
View return policy
Description
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.Specifications
| Galectin-1 | |
| Polyclonal | |
| Unconjugated | |
| LGALS1 | |
| 14 kDa laminin binding protein, 14 kDa lectin, Gal 1, GAL1, Galaptin, Galectin 1, Galectin-1, HBL, HLBP14, HPL, Lactose binding lectin 1, LGALS1, S Lac lectin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16852, 3956, 56646 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09382, P11762, P16045 | |
| Lgals1 | |
| Galectin-1 Fusion Protein Ag2432 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction