missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59002
This item is not returnable.
View return policy
Description
Galectin-1 Polyclonal specifically detects Galectin-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Galectin-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected to cross react based on sequence identity: Rabbit: 86%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Sheep | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml | |
| P09382 | |
| LGALS1 | |
| Synthetic peptides corresponding to LGALS1 (lectin, galactoside-binding, soluble, 1 (galectin-1)) The peptide sequence was selected from the middle region of LGALS1. Peptide sequence EQREAVFPFQPGSVAEVCITFDQANLTVKLPDGYEFKFPNRLNLEAINYM. | |
| 100 μL | |
| Apoptosis | |
| 3956 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction