missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-1 Antibody (GAL1/2499R), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-79855UV
This item is not returnable.
View return policy
Description
Galectin-1 Monoclonal specifically detects Galectin-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Galectin-1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LGALS1 | |
| Recombinant fragment (around aa 12-108) of human Galectin-1 protein (exact sequence is proprietary) (Uniprot: P09382) | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins | |
| Human | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GAL1/2499R | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction