missing translation for 'onlineSavingsMsg'
Learn More

Galectin-1 Antibody (001), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18336941 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336941 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336941 Supplier Novus Biologicals Supplier No. NBP289387AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Galectin-1 Monoclonal specifically detects Galectin-1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-1
Applications Western Blot
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Galectin-1 (NP_002296.1; Met1-Asp135).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 3956
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.