missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572315
This item is not returnable.
View return policy
Description
GALC Polyclonal Antibody for Western Blot, Flow
This gene encodes a lysosomal protein which hydrolyzes the galactose ester bonds of galactosylceramide, galactosylsphingosine, lactosylceramide, and monogalactosyldiglyceride. Mutations in this gene have been associated with Krabbe disease, also known as globoid cell leukodystrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| GALC | |
| Polyclonal | |
| Unconjugated | |
| Galc | |
| 2310068B06Rik; 3.2.1.46; A930008M05Rik; AW212969; AW413532; beta-galactosidase; casein kinase 2, beta polypeptide pseudogene; Gacy; Galactocerebrosidase; galactocerebroside beta-galactosidase; Galactosylceramidase; galactosylceramidase (Krabbe disease); galactosylceramide beta-galactosidase; galactosylceraminidase; GALC; GALCERase; testis tissue sperm-binding protein Li 88E; testis tissue sperm-binding protein Li 89A; twi; twitcher | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 2581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P54803 | |
| Galc | |
| KLH conjugated synthetic peptide between 26-56 amino acids from human GALC. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction