missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galanthamine Hydrobromide, TRC
Specifications
Specifications
| Chemical Name or Material | Galantamine Hydrobromide |
| CAS | 1953-04-4 |
| Molecular Formula | C17 H21 N O3 . Br H |
| Quantity | 50 mg |
| Synonym | 6H-Benzofuro[3a,3,2-ef][2]benzazepin-6-ol, 4a,5,9,10,11,12-hexahydro-3-methoxy-11-methyl-, hydrobromide (1:1), (4aS,6R,8aS)-, 6H-Benzofuro[3a,3,2-ef][2]benzazepin-6-ol, 4a,5,9,10,11,12-hexahydro-3-methoxy-11-methyl-, hydrobromide, (4aS,6R,8aS)- (9CI), Galanthamine, hydrobromide (6CI,8CI), (-)-Galantamine hydrobromide, (-)-Galanthamine hydrobromide, Galantamine hydrobromide, Galanthamine hydrogen bromide, Jilkon hydrobromide, Lycoremine hydrobromide, Nivalin, Nivaline, Nivaline (pharmaceutical), Razadyne, Reminyl, Tamilin, (4aS,6R,8aS)-3-Methoxy-11-methyl-5,6,9,10,11,12-hexahydro-4aH-[1]benzofuro[3a,3,2-ef][2]benzazepin-6-ol hydrobromide |
| InChI Formula | InChI=1S/C17H21NO3.BrH/c1-18-8-7-17-6-5-12(19)9-14(17)21-16-13(20-2)4-3-11(10-18)15(16)17;/h3-6,12,14,19H,7-10H2,1-2H3;1H/t12-,14-,17-;/m0./s1 |
| SMILES | Br.COc1ccc2CN(C)CC[C@@]34C=C[C@H](O)C[C@@H]3Oc1c24 |
| IUPAC Name | (4aS,6R,8aS)-4a,5,9,10,11,12-Hexahydro-3-methoxy-11-methyl-6H-benzofuro[3a,3,2-ef][2]benzazepin-6-ol hydrobromide |
| Molecular Weight (g/mol) | 368.27 |
| Formula Weight | 367.0783 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?