missing translation for 'onlineSavingsMsg'
Learn More

Galanthamine hydrobromide, MedChemExpress

Product Code. 16417986
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16417986

Brand: Medchem Express HYA0009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Galanthamine hydrobromide (Galantamine hydrobromide) is a selective, reversible, competitive, alkaloid AChE inhibitor, with an IC50 of 0.35 μM. Galanthamine hydrobromide is a potent allosteric potentiating ligand (APL) of human α3β4, α4β2, α6β4 nicotinic receptors ( nAChRs). Galanthamine hydrobromide is developed for the research of Alzheimer's disease (AD).

Galanthamine hydrobromide (Galantamine hydrobromide) is a selective, reversible, competitive, alkaloid AChE inhibitor, with an IC50 of 0.35 μM. Galanthamine hydrobromide is a potent allosteric potentiating ligand (APL) of human α3β4, α4β2, α6β4 nicotinic receptors ( nAChRs). Galanthamine hydrobromide is developed for the research of Alzheimer's disease (AD).

Specifications

Chemical Name or Material Galanthamine hydrobromide
CAS 4-4-1953
Color White
Purity Grade Notes Research
Molecular Formula C17H22BrNO3
Quantity 100 mg
Synonym Galantamine hydrobromide
Solubility Information H2O : 16.67 mg/mL (45.27 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : ≥ 3.7 mg/mL (10.05 mM)
SMILES O[C@@H]1C[C@@H]2OC3=C4C(CN(C)CC[C@]42C=C1)=CC=C3OC.Br
Molecular Weight (g/mol) 368.27
Formula Weight 368.27
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.