Learn More
Galanin R2/GALR2 Antibody (616520), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB6544PECY7
Product Code. 30029169
  • £476.00 / 0.10mL
Description
Galanin R2/GALR2 Monoclonal antibody specifically detects Galanin R2/GALR2 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galanin R2/GALR2
Monoclonal
PE-Cyanine7
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry
616520
Flow Cytometry
GAL2-R, galanin receptor 2, galanin receptor type 2, GALNR2MGC125984, GALR-2, MGC125983
NS0 mouse myeloma cell line transfected with human Galanin R2, Accession # O43603
0.1 mL
GPCR
8811
Store at 4C in the dark. Do not freeze.
IgG2b