Learn More
Invitrogen™ Galacton-Star™ Substrate (50X Concentrate)
Concentrated chemiluminescent substrate for E.coli β-galactosidase assays. Thermo Scientific Galacton-Star Substrate (50X Concentrate) is highly sensitive which makes it ideal for detection of weak expression and for transfection normalization with o
Brand: Invitrogen™ T2265
Code : NEW
Additional Details : Weight : 0.02000kg
Description
Galacton-Star substrate is a chemiluminescent substrate designed for the rapid, and sensitive detection of beta-galactosidase in cell lysates. The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric beta-galactosidase assays. Light emission from Galacton-Star reaches maximum in 60-90 minutes and remains constant for at least 1 hour. After incubation at room temperature, the signal is measured in a luminometer. As few as 10 femtograms of beta-galactosidase (20,000 molecules) are detectable.Specifications
ELISA Substrate | |
Chemiluminescent | |
Bottle | |
Beta-Gal | |
Room Temperature |
4 mL | |
Liquid | |
Galacton-Star™, NovaBright™ | |
Store the product at 4°C. |
For Research Use Only. Not for use in diagnostic procedures.