missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Galacton-Plus™ Substrate (100X Concentrate)

Product Code. 12694315
Click to view available options
Quantity:
2mL
4mL
10mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12694315

Brand: Molecular Probes™ T2118

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Galacton-Plus™ Substrate (100X Concentrate)

The Galacton-Plus™ Substrate is a concentrated chemiluminescent substrate for E.coli β-galactosidase assays.

For Research Use Only. Not for use in diagnostics procedures.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the product at 4°C.
Form Liquid
Product Line Galacton-Plus™
Quantity 2mL
Substrate Beta-Galactosidase
Substrate Type Beta-Gal Substrate
Format Bottle(s)
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.