missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galacto-Star™ β-Galactosidase Reporter Gene Assay System for Mammalian Cells
The Galacto-Star™ One-Step β-Galactosidase Reporter Gene Assay System chemiluminescent reporter gene assay system enables rapid, sensitive detection of β-galactosidase in mammalian cell lysates. SensitiveThe chemiluminescent assay exhibits over three o
£306.00 - £668.00
Specifications
Product Type | β-Galactosidase Reporter Gene Assay System |
---|---|
Detection Method | Chemiluminescent |
For Use With (Equipment) | Luminometer (Microplate) |
Assay | β-Gal (lacZ) Assay |
Content And Storage | This kit contains the lysis solution, substrate, enhancer, and required buffer. Store the product at 4°C. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12624275
|
Invitrogen™
T1012 |
600 Assays |
£306.00
Each |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12644275
|
Invitrogen™
T1014 |
1800 Assays |
£668.00
Each |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Glow assay kinetics provides a “window” during which measurements may be performed, facilitating HTS applications where assay automation is used
- Can be used with either mammalian or yeast model systems, providing flexibility in choice of model systems
- Wide dynamic range of β-galactosidase assay lets the user measure enzyme level accurately from femtogram to nanogram range
- Assay sensitivity is 100- to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assays
- Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
- Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
- Assay can be completed in about one hour, providing fast assay turnaround
Sensitive
The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity than colorimetric β-galactosidase assays and can be completed in a fraction of the time required for assays for chloramphenicol acetyltransferase (CAT) and ELISA-based reporter assays.
Versatile
The Galacto-Star system includes Galacton-Star chemiluminescent substrate for β-galactosidase and is suited for measurement in both luminometers and scintillation counters. Automatic injection is not required.
Long-Lasting Signal
Light emission from the Galacto-Star assay reaches maximum in 60 to 90 minutes and remains constant for nearly one hour. Cell lysate is mixed with reaction buffer containing Galacton-Star substrate and Sapphire-II™ luminescence enhancer. After a 60-90 minute incubation at room temperature, the signal is measured in a luminometer or scintillation counter.
Specifications
- Assay Type: β-Gal (lacZ) Assay
- Compatible Cells: Mammalian Cells
- Detection Method: Chemiluminescent
- For Use With Equipment: Luminometer (Microplate)
- Form: Liquid
- Format: 96-well plate
- High Throughput Compatibility: High Throughput-Compatible
- Readout: End Point
- Sample Type (Specific): Cell Cultures
- Substrate: Galacton-Star
- Substrate Properties: Chemical Substrate
- Substrate Type: Beta-Gal Substrate
- Target Enzyme: Beta-Galactosidase
Specifications
β-Galactosidase Reporter Gene Assay System | |
Luminometer (Microplate) | |
This kit contains the lysis solution, substrate, enhancer, and required buffer. Store the product at 4°C. | |
High-throughput Compatible | |
End Point | |
Galacton-Star™ | |
Chemical Substrate | |
Galacto-Star™, NovaBright™ | |
Liquid |
Chemiluminescent | |
β-Gal (lacZ) Assay | |
Mammalian Cells | |
Unconjugated | |
Cell Cultures | |
Beta-Gal Substrate | |
Beta-Galactosidase | |
Enzyme Assay |
For Research Use Only. Not for use in diagnostic procedures.