missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galactaric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Mucic acid |
| CAS | 526-99-8 |
| Molecular Formula | C6 H10 O8 |
| Quantity | 500 g |
| Synonym | Galactaric acid, Galactosaccharic acid, Saccharolactic acid |
| InChI Formula | InChI=1S/C6H10O8/c7-1(3(9)5(11)12)2(8)4(10)6(13)14/h1-4,7-10H,(H,11,12)(H,13,14) |
| SMILES | OC(C(O)C(O)C(=O)O)C(O)C(=O)O |
| IUPAC Name | 2,3,4,5-tetrahydroxyhexanedioic acid |
| Molecular Weight (g/mol) | 210.14 |
| Formula Weight | 210.0376 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?