missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAL3ST2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564472
This item is not returnable.
View return policy
Description
Immunogen sequence: YAKNNMWFDF GFDPNAQCEE GYVRARIAEV ERRFRLVLIA EHLDESLVLL Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 68%.
Specifications
| GAL3ST2 | |
| Polyclonal | |
| Unconjugated | |
| Gal3st2 | |
| Beta-galactose-3-O-sulfotransferase 2; gal 3-O-sulphotransferase; GAL3ST2; Gal3ST-2; Galactose-3-O-sulfotransferase 2; gal-beta-1, 3-GalNAc 3'-sulfotransferase 2; galbeta1-3GalNAc 3'-sulfotransferase 2; glycoprotein beta-Gal 3'-sulfotransferase 2; GP3ST | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64090 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H3Q3 | |
| Gal3st2 | |
| Recombinant protein corresponding to Human GAL3ST2. Recombinant protein control fragment (Product #RP-104009). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction