missing translation for 'onlineSavingsMsg'
Learn More

GAL3ST1 Antibody, Novus Biologicals™

Product Code. 18349958 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349958 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349958 Supplier Novus Biologicals Supplier No. H00009514D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GAL3ST1 Polyclonal antibody specifically detects GAL3ST1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAL3ST1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH14649.1
Gene Alias 3'-phosphoadenosine-5'-phosphosulfate:GalCer sulfotransferase, 3'-phosphoadenylylsulfate:galactosylceramide 3'-sulfotransferase, cerebroside (3'-phosphoadenylylsulfate:galactosylceramide 3') sulfotransferase, CSTCerebroside sulfotransferase, EC 2.8.2.11, galactose-3-O-sulfotransferase 1, galactosylceramide sulfotransferase, GalCer sulfotransferase
Host Species Rabbit
Immunogen GAL3ST1 (AAH14649.1, 1 a.a. - 423 a.a.) full-length human protein. MLPPQKKPWESMAKGLVLGALFTSFLLLMYSYAVPPLHAGLASTTPEAAASCSPPALEPEAVIRANGSAGECQPRRNIVFLKTHKTASSTLLNILFRFGQKHRLKFAFPNGRNDFDYPTFFARSLVQDYRPGACFNIICNHMRFHYDEVRGLVPTNAIFITVLRDPARLFESSFHYFGPVVPLTWKLSAGDKLTEFLQDPDRYYDPNGFNAHYLRNLLFFDLGYDNSLDPSSPQVQEHILEVERRFHLVLLQEYFDESLVLLKDLLCWELEDVLYFKLNARRDSPVPRLSGELYGRATAWNMLDSHLYRHFNASFWRKVEAFGRERMAREVAALRHANERMRTICIDGGHAVDAAAIQDEAMQPWQPLGTKSILGYNLKKSIGQRHAQLCRRMLTPEIQYLMDLGANLWVTKLWKFIRDFLRW
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9514
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.