missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gal-Screen™ β-Galactosidase Reporter Gene Assay System for Mammalian Cells

Product Code. 12674285
Click to view available options
Quantity:
10,000 Assays
1000 Assays
200 Assays
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12674285

Brand: Invitrogen™ T1029

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Enzyme substrate and reaction buffer

Combines direct cell lysis with rapid, ultra-sensitive chemiluminescent detection of β-galactosidase reporter enzyme.

  • Homogeneous assay format allows detection of β-galactosidase in presence of normal culture media without removal of media and without an additional cell lysis step, providing the easiest and most streamlined assay procedure possible
  • The kinetics of glow-assay provides a window during which measurements may be performed, facilitating HTS applications where assay automation is used
  • Can be used with either mammalian or yeast model systems, providing flexibility in choice of model systems
  • Wide dynamic range of β-galactosidase assay enables accurate measurement of enzyme from femtogram to nanogram range
  • Assay sensitivity is 100 to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assay technologies
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
  • Assay can be completed in about one hour, providing fast assay turnaround

Ideal for Screening
This homogeneous assay is ideally suited for screening applications where assay automation is required. The Gal-Screen system uses Galacton-Star™ chemiluminescent substrate for convenient measurement in a luminometer. Light emission reaches maximum in 60-90 minutes and remains constant for 45-90 minutes.

Specifications

  • Assay Type: β-Gal (lacZ) Assay
  • Compatible Cells: Mammalian Cells
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Form: Liquid
  • Format: 96-well plate
  • Readout: End Point
  • Sample Type (Specific): Cell Cultures
  • Substrate: Galacton-Star™
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Beta-Gal Substrate
  • Target Enzyme: Beta-Galactosidase
  • Technique: Enhanced Chemiluminescence
TRUSTED_SUSTAINABILITY

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Detection Method Chemiluminescence
For Use With (Equipment) Luminometer (Microplate)
Assay β-Gal (lacZ) Assay
Content And Storage This kit contains the enzyme substrate and the reaction buffer.
Store the product at 4°C.
Compatible Cells Mammalian Cells
High-throughput Compatibility High-throughput Compatible
Label or Dye Unconjugated
Readout End Point
Sample Type Cell Cultures
Substrate Galacton-Star™
Substrate Type Beta-Gal Substrate
Substrate Properties Chemical Substrate
Target Enzyme Beta-Galactosidase
Product Line Gal-Screen, NovaBright
For Use With (Application) Enzyme Assay
Quantity 200 Assays
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.