missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599201
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GAK.
GAK serine/threonine kinase or Cyclin-G-associated kinase was originally identified as a protein that is associated with cyclin G. However, neither CDK5 nor GAK kinase activity is activated by cyclin G nor does GAK affect CDK5 kinase activity. GAK seems to act as an auxilin homolog that is involved in the uncoating of clathrin-coated vesicles by Hsc70 in non-neuronal cells. Expression oscillates slightly during the cell cycle, peaking at G1.
Specifications
| GAK | |
| Polyclonal | |
| Unconjugated | |
| GAK | |
| auxilin-2; cyclin G associated kinase; cyclin G-associated kinase; cyclin-G-associated kinase; D130045N16Rik; DNAJ26; DNAJC26; GAK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 231580, 2580 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14976, Q99KY4 | |
| GAK | |
| A synthesized peptide derived from human GAK(Accession O14976), corresponding to amino acid residues K109-V159. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction